Order Entry
Ireland
ContactUsLinkComponent
Anti-MLF1IP Rabbit Polyclonal Antibody
Anti-MLF1IP Rabbit Polyclonal Antibody
Catalog # PRSI49-359
Supplier:  ProSci Inc.
undefined
Anti-MLF1IP Rabbit Polyclonal Antibody
Catalog # PRSI49-359
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    MLF1 interacting Protein
  • Antigen symbol:
    MLF1IP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Chimpanzee,
    Human,
    Bovine,
    Dog
  • Western blot:
    Yes
  • Form:
    Liquid
  • Immunogen:
    MLF1IP antibody was raised against amino acids surrounding Thr78 of MLF1IP (Human).
  • Tested applications:
    Immunoaffinity Chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Myeloid leukemia factor-1 (MLF1) Interacting Protein (also known as PBIP1, MLF1IP1, KLIP1 or KSHV latent nuclear antigen interacting protein 1) is a novel polo-like kinase 1 (Plk1) substrate. Plk1 phosphorylation of MLF1IP induces ubiquitination and degradation of MLF1IP prior to the metaphase/anaphase transition. Several Plk1-dependent phosphorylation sites have been identified on MLF1IP by mass spectrometry. Mutations of these sites stabilize MLF1IP and inhibit mitotic progression. Subsequent in vitro and in vivo MLF1IP phosphorylation and stability assays have revealed that phosphorylation of Thr78 is critical for triggering Plk1-dependent MLF1IP degradation. Expression of a non-degradable Thr78Ala mutant was sufficient to induce a mitotic block. Timely phosphorylation of MLF1IP on Thr78 by Plk1 is critical for eliminating the MLF1IP-imposed mitotic block prior to anaphase onset. MLF1IP is speculated to be a novel tumor suppressor, whose function is required for proper sister-chromatid separation. Loss of MLF1IP function may result in improper segregation of chromosomes and genomic instability, thus promoting tumorigenesis.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MLF1IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Human