Anti-FOXC2 Goat Polyclonal Antibody
Catalog # PRSI45-605
Supplier: ProSci Inc.
Specifications
- Antigen name:Forkhead Box C2
- Antigen symbol:FOXC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MFH1|MFH-1|FKHL14|LD
- Concentration:500 ug/mL
- Immunogen:FOXC2 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of FOXC2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
FOXC2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
FOXC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FOXC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human