Anti-EGLN3 Goat Polyclonal Antibody
PRSI45-520
- Antigen name:Egl-9 Family Hypoxia-Inducible Factor 3
- Antigen symbol:EGLN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:egl nine homolog 3|MGC125999|egl nine-like protein 3 isoform|MGC125998|EGL nine (C.elegans) homolog 3|PHD3|EGLN3 |HIFPH3|egl nine homolog 3 (C. elegans) |FLJ21620
- Concentration:500 ug/mL
- Immunogen:EGLN3 antibody was raised against a 12 amino acid synthetic peptide near the internal region of EGLN3.
- Size:100 µg
- Pk:100 µG
EGLN3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
EGLN3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EGLN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human