Anti-ARHGAP22 Rabbit Polyclonal Antibody
Catalog # PRSI39-854
Supplier: ProSci Inc.
Specifications
- Antigen name:Rho GTPase Activating Protein 22
- Antigen symbol:ARHGAP22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:ARHGAP22|Rho-type GTPase-activating protein 22|RHOGAP2|Rho GTPase-activating protein 22
- Immunogen:A synthetic peptide corresponding to the region surrounding pS397.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Rho GTPase-activating protein 22 involved in the signal transduction pathway that has been shown to be important during capillary formation during angiogenesis. It is also an activator for the RAC1.
Affinity Purified
Type:
Antigen: ARHGAP22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat