Anti-Clostridum difficile Mouse Monoclonal Antibody
Catalog # PRSI35-601
Supplier: ProSci Inc.
Specifications
- Antigen symbol:Clostridum difficile
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Bacteria
- Form:Liquid
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Clostridum difficile toxin antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.
Clostridum difficile toxin antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen:
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: