Anti-AdV Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI35-491
Supplier: ProSci Inc.
Specifications
- Antigen name:Adenovirus antigen
- Antigen symbol:AdV
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Virus
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:AdV|Adeno Virus|HADV
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Adenovirus antibody is conjugated with high purity isomer I of fluorescein isothiocyanate. Care is taken to ensure complete removal of any free fluorescein from the final product.
Adenovirus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AdV
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus