Order Entry
Ireland
ContactUsLinkComponent
Anti-RFC3 Rabbit Polyclonal Antibody
Anti-RFC3 Rabbit Polyclonal Antibody
Catalog # PRSI30-152
Supplier:  ProSci Inc.
undefined
Anti-RFC3 Rabbit Polyclonal Antibody
Catalog # PRSI30-152
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Replication factor C subunit 3
  • Antigen symbol:
    RFC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Dog
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Antigen synonyms:
    RFC38
  • Immunogen:
    A synthetic peptide corresponding a region of human RFC3.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. RFC3 is the 38 kDa subunit. This subunit is essential for the interaction between the 140 kDa subunit and the core complex that consists of the 36, 37, and 40 kDa subunits.The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 38 kD subunit. This subunit is essential for the interaction between the 140 kD subunit and the core complex that consists of the 36, 37, and 40 kD subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RFC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human