Anti-PSG1 Rabbit Polyclonal Antibody
Catalog # PRSI29-736
Supplier: ProSci Inc.
Specifications
- Antigen name:Pregnancy Specific Beta-1-glycoprotein 1
- Antigen symbol:PSG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human PSG1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The function remains unknown.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PSG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human