Anti-KIF1C Rabbit Polyclonal Antibody
Catalog # PRSI27-977
Supplier: ProSci Inc.
Specifications
- Antigen name:Kinesin Family member 1C
- Antigen symbol:KIF1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:SPAX2|LTXS1|SATX2|SPG58|SAX2
- Immunogen:A synthetic peptide corresponding a region of human KIF1C.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
KIF1C represents a member of the Unc104 subfamily of kinesin-like proteins that are involved in the transport of mitochondria or synaptic vesicles in axons. KIF1C consists of an amino-terminal motor domain followed by a U104 domain and probably binds to target membranes through carboxyl-terminal sequences.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KIF1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human