Anti-MLLT3 Rabbit Polyclonal Antibody
PRSI25-437
- Antigen name:Myeloid/lymphoid or Mixed-lineage Leukemia (trithorax homolog Drosophila); translocated to 3
- Antigen symbol:MLLT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Myeloid/lymphoid or mixed-lineage leukemia translocated to chromosome 3 protein|AF9|YEATS3|ALL1 fused gene from chromosome 9 protein
- Immunogen:A synthetic peptide corresponding a region of human MLLT3.
- Size:50 µg
- Pk:50 µG
MLLT3 is a regulator of early erythroid and megakaryocytic cell fate in the human system. Expression of MLLT3 in human CD34+ cells induces acute myeloid, lymphoid, or mixed-lineage leukemia in immunodeficient mice. Translocation t (9;11) (p22;q23), a chromosomal aberration involving MLLT3 is associated with acute leukemias.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MLLT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse