Order Entry
Ireland
ContactUsLinkComponent
Anti-CHD2 Rabbit Polyclonal Antibody
Anti-CHD2 Rabbit Polyclonal Antibody
Catalog # PRSI25-199
Supplier:  ProSci Inc.
undefined
Anti-CHD2 Rabbit Polyclonal Antibody
Catalog # PRSI25-199
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    ChromoDomain Helicase DNA Binding Protein 2
  • Antigen symbol:
    CHD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    EEOC
  • Immunogen:
    A synthetic peptide corresponding a region of human CHD2.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template.The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

CHD2 antibody can be used for detection of CHD2 by ELISA at 1:62500. CHD2 antibody can be used for detection of CHD2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHD2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: