Anti-SLAIN1 Rabbit Polyclonal Antibody
PRSI5467
- Antigen name:SLAIN Motif Family Member 1
- Antigen symbol:SLAIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:SLAIN motif-containing protein 1|C13orf32
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:SLAIN1 antibody was raised against a 16 amino acid synthetic peptide near the center of human SLAIN1.
- Tested applications:E, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
SLAIN1 Antibody: SLAIN1 is a novel stem cell protein that was identified by transciptional profiling of mouse and human embryonic stem cells (ESCs) and is expressed at the stem cell and epiblast stages. Unlike its more widely expressed homolog SLAIN2, SLAIN1 was found to be expressed in the epiblast, nervous system, tailbud and somites of the developing mouse embryo later experiments suggested that SLAIN1 may play a role in the development of the nervous system as well as in the morphogenesis of several embryonic structures.
Type:
Antigen: SLAIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human