Order Entry
Ireland
ContactUsLinkComponent
Anti-ZIP8 Rabbit Polyclonal Antibody
Anti-ZIP8 Rabbit Polyclonal Antibody
  PRSI6095
undefined
Anti-ZIP8 Rabbit Polyclonal Antibody
  PRSI6095

 

  • Antigen symbol:
    ZIP8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    ZIP8 antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human ZIP8.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

ZIP8 Antibody: The zinc transporter ZIP8, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP8 is glycosylated and located at the plasma membrane and mitochondria. It has been identified as the transporter responsible for transport of the toxic Cadmium cation. ZIP8 has also been suggested to play a role in the regulation of interferon-gamma expression in activated human T cells.

Type:
Antigen: ZIP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human