Supelco® Drug and metabolite standards, prednisone, LoGiCal®
Catalog # PROOLGCFOR0482.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:53-03-2
- Description:Prednisone
- Formula:C₂₁H₂₆O₅
- Description:Prednisone