Drug and metabolite standards, hyoscine hydrobromide trihydrate, LoGiCal®
Catalog # PROOLGCFOR0546.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:6533-68-2
- Description:Hyoscine hydrobromide trihydrate
- Formula:C₁₇H₂₈BrNO₇
- Description:Hyoscine hydrobromide trihydrate