Drug and metabolite standards, desmethylclomipramine hydrochloride, LoGiCal®
Catalog # PROOLGCFOR0545.08
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:29854-14-6
- Description:Desmethylclomipramine hydrochloride
- Formula:C₁₈H₂₂Cl₂N₂
- Description:Desmethylclomipramine hydrochloride