Order Entry
Ireland
ContactUsLinkComponent
Drug and metabolite standards, chlortetracycline hydrochloride, LoGiCal®
Drug and metabolite standards, chlortetracycline hydrochloride, LoGiCal®
Catalog # PROOLGCFOR0607.00
CAS Number:  
Drug and metabolite standards, chlortetracycline hydrochloride, LoGiCal®
Catalog # PROOLGCFOR0607.00
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    64-72-2
  • Description:
    Chlortetracycline hydrochloride
  • Formula:
    C₂₂H₂₄Cl₂N₂O₈
  • Description:
    Chlortetracycline hydrochloride

Specifications

About this item