Order Entry
Ireland
ContactUsLinkComponent
 
Pyrantel Related Compounds, Reference Standard
 :  USP
 
Pyrantel Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    33401-94-4
  • UN No.:
    2811
  • Formula:
    C₁₅H₂₀N₂O₆S
  • Description:
    Pyrantel Tartrate
  • Synonym(s):
    (Z)-1-Methyl-2-[2-(2-thiophenyl)vinyl]-1,4,5,6-Tetrahydropyrimidine 4,4'-methylenebis[3-hydroxy-2-naphthoate] (1:1)

 

Frequently Bought Together