Anti-AGRP Rabbit Polyclonal Antibody
BSBTA00272-1
- Antibody type:Primary
- Antigen symbol:AGRP
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for AGRP detection. Tested with IHC-P, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: AGRP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat