Anti-Laminin Rabbit Polyclonal Antibody
Catalog # BSBTA03522-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Laminin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Laminin detection. Tested with WB, IHC-P in Human.
Type: Primary
Antigen: Laminin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human