Anti-Neurocan Rabbit Polyclonal Antibody
Catalog # BSBTA06700-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Neurocan
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Neurocan detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: Neurocan
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat