Anti-IL17F Rabbit Polyclonal Antibody
BSBTA02062-1
- Antibody type:Primary
- Antigen symbol:IL17F
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for IL17F detection. Tested with WB, Direct ELISA in Human;Rat.
Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat