Anti-IL1 beta Rabbit Polyclonal Antibody
Catalog # BSBTA00101-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:IL1 beta
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for IL1 beta detection. Tested with WB in Mouse;Rat.
Type: Primary
Antigen: IL1 beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat