Anti-SIX3 Rabbit Polyclonal Antibody
BSBTA02604
- Antibody type:Primary
- Antigen name:Homeobox protein SIX3
- Antigen symbol:SIX3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Homeobox protein SIX3(SIX3) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: SIX3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat