Anti-NOV Rabbit Polyclonal Antibody
Catalog # BSBTA06319-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Protein NOV homolog
- Antigen symbol:NOV
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Protein NOV homolog(NOV) detection. Tested with WB, IHC-P in Human;Rat.
Type: Primary
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat