Order Entry
Ireland
ContactUsLinkComponent
Pamidronate Disodium, Reference Standard
Pamidronate Disodium, Reference Standard
Catalog # 1492801.
Supplier:  USP
CAS Number:  
Pamidronate Disodium, Reference Standard
Catalog # 1492801.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    USP reference standard
  • CAS No.:
    109552-15-0
  • Description:
    Pamidronate Disodium
  • Formula:
    C₃H₁₉NNa₂O₁₂P₂
  • Description:
    Pamidronate Disodium

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard