Anti-IGF1 Receptor Rabbit Polyclonal Antibody
BSBTA00070
- Antibody type:Primary
- Antigen name:IGF1 Receptor
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for IGF1 Receptor detection. Tested with WB, IHC-P, Direct ELISA in Mouse;Rat.
Type: Primary
Antigen: IGF1 Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat