Anti-Kallikrein 6 Rabbit Polyclonal Antibody
BSBTA01882-2
- Antibody type:Primary
- Antigen name:Kallikrein 6
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Kallikrein 6 detection. Tested with WB, Direct ELISA in Human.
Type: Primary
Antigen: Kallikrein 6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human