Order Entry
Ireland
Orders LinkContactUsLinkComponent
Thiostrepton, Millipore®

Thiostrepton, Millipore®

Thiostrepton, Millipore®
Certificates
Thiostrepton, Millipore®

2 Options Available Below

Product Details & Documents

Thiostrepton is a thiazole-containing peptide antibiotic that inhibits protein synthesis by preventing binding of GTP to 50S ribosomal subunit.

Thiostrepton inhibits the function of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome. The thiostrepton-resistant gene is also commonly used as a selective marker for recombinant DNA/plasmid technologies.

Synonyms:  N-[(7R,8S)-2-Carboxy-7,8-dihydro-8-hydroxy-4-[(1S)-1-hydroxyethyl]-7-quinolinyl]-L-isoleucyl-L-alanyl-2,3-didehydroalanyl-L-alanyl-2-[(4aR,11S,14Z,18S,21S,28S,32aS)-4a-amino-21-[(1S,2R)-1,2-dihydroxy-1-methylpropyl]-14-ethylidene-3,4,4a,9,10,11,12,13,14,18,19,20,21,27,28,32a-hexadecahydro-11,28-bis[(1R)-1-hydroxyethyl]-9,12,19,26-tetraoxo-17H,26H-8,5:18,15:25,22:32,29-tetranitrilo-5H,15H-pyrido[3,2-m][1,11,17,24,4,7,20,27]tetrathiatetraazacyclotriacontin-2-yl]-4-thiazolecarbonyl-2,3-didehydroalanyl-2,3-didehydro-alaninamide,(1→528)-lactone
  • CAS Number:1393-48-2
  • Formula:C₇₂H₈₅N₁₉O₁₈S₅
  • MDL Number:MFCD00135828
  • Molecular Weight:1664.887 g/mol
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 141,00 €
    141,00 €
    1 * 1 g
       141,00 €

  • 10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 1 130,00 €
    1 130,00 €
    1 * 10 g
       1 130,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.