δ-methrin ≥98% (by TLC), white solid, Calbiochem®
Catalog # 253300-10
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
- Environmentally Preferable:
Specifications
About this item
Synonyms: (S)-alpha-Cyano-3-phenoxybenzyl (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylate, (1R,3R)-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylic acid (S)-alpha-Cyano-3-phenoxybenzyl ester, Deltamethrin
:
:505.206 g/mol
UN:2811
:GHS05
:GHS09
Storage Temperature:Refrigerator
CAS Number:52918-63-5
Formula:C₂₂H₁₉Br₂NO₃
MDL Number:MFCD00870122
ADR:6.1 II