(-)-Indolactam V ≥97%, solid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:90365-57-4
:
Formula:C₁₇H₂₃N₃O₂
:301.39 g/mol
Specification Test Results
- Formal name:(2S,5S)-1,2,4,5,6,8-hexahydro-5-(hydroxymethyl)-1-methyl-2-(1-methylethyl)-3H-pyrrolo[4,3,2-gh]-1,4-benzodiazonin-3-one
- CAS number:90365-57-4
- Molecular formula:C17H23N3O2
- Formula weight:301,4
- Purity:≥97%
- Formulation:A white to off-white solid
- λmax:203, 229 and 301 nm
- Storage:4 °C
- Stability:≥ 2 years
Specification Test Results
Frequently Bought Together