Fmoc-NH-(PEG)₂-COOH (20 atoms) ≥92% (by HPLC), Novabiochem®
:916585-44-9
:
:C₂₉H₃₈N₂O₉
:MFCD17019373
:558.62 g/mol
- Environmentally Preferable:
- Colour:White to slight yellow to beige
- Appearance of substance:Powder
- Identity (IR):Passes test
- Purity (TLC(0110)):≥ 92%
- Assay (HPLC, area %):≥ 92.0% (a/a)
- Storage:Store at 2...8 °C
Frequently Bought Together