Order Entry
Ireland
ContactUsLinkComponent
Indium standard solution
Indium standard solution
Catalog # PROONIST-3124A
CAS Number:  
Indium standard solution
Catalog # PROONIST-3124A
CAS Number:  
Specifications
  • Pk:
    5
  • Description:
    NIST-3124A Indium standard solution
  • Element:
    Indium
  • Symbol:
    In
Specifications
About this item
This reference standard is intended for use as a primary calibration standard for the quantitative determination of indium.
A unit consists of five 10 ml sealed borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of indium.
Certifications: Traceable to NIST and SRM numbers.