Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, tetra-O-acetyl-mannose triflate, Ph. Eur. RS
Organic reference standards, tetra-O-acetyl-mannose triflate, Ph. Eur. RS
Catalog # EDQMY0001087
Supplier:  EDQM
CAS Number:  
Organic reference standards, tetra-O-acetyl-mannose triflate, Ph. Eur. RS
Catalog # EDQMY0001087
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    60 mg
  • Standards:
    Reference standard
  • CAS No.:
    92051-23-5
  • Description:
    Tetra-O-acetyl-mannose triflate
  • Formula:
    C₁₅H₁₉F₃O₁₂S
  • Description:
    Tetra-O-acetyl-mannose triflate

Specifications

About this item