Order Entry
Ireland
ContactUsLinkComponent
Impurity and API standard, tenofovir disoproxil fumarate
Impurity and API standard, tenofovir disoproxil fumarate
Catalog # PROOMM1329.00
CAS Number:  
Impurity and API standard, tenofovir disoproxil fumarate
Catalog # PROOMM1329.00
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    202138-50-9
  • Description:
    Tenofovir disoproxil fumarate
  • Formula:
    C₂₃H₃₄N₅O₁₄P
  • Description:
    Tenofovir disoproxil fumarate

Specifications

About this item