Order Entry
Ireland
ContactUsLinkComponent
Impurity and API standard, ritalinic acid
Impurity and API standard, ritalinic acid
Catalog # PROOMM1031.04
CAS Number:  
Impurity and API standard, ritalinic acid
Catalog # PROOMM1031.04
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    19395-41-6
  • Description:
    Ritalinic acid
  • Formula:
    C₁₃H₁₇NO₂
  • Description:
    Ritalinic acid

Specifications

About this item