Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standards, rivastigmine, Ph. Eur., RS
 :  EDQM
 
Organic reference standards, rivastigmine, Ph. Eur., RS
 :  EDQM

 

  • CAS No.:
    415973-05-6
  • Formula:
    C₁₄H₂₂N₂O₂
  • Description:
    Rivastigmine impurity D, 3-[(1R)-1-(dimethylamino)ethyl]phenyl ethyl(methyl)carbamate ((R)-enantiomer)

 

Frequently Bought Together