Order Entry
Ireland
ContactUsLinkComponent
 
Impurity and API standard, 2-bromo-3'-chloropropiophenone
  PROOMM0745.15
 :  
Impurity and API standard, 2-bromo-3'-chloropropiophenone
  PROOMM0745.15
 :  

 

  • Pk:
    1
  • Standards:
    Reference standards
  • CAS No.:
    34911-51-8
  • Description:
    2-Bromo-1-(3-chlorophenyl)propan-1-one
  • Formula:
    C₉H₈BrClO
  • Description:
    2-Bromo-1-(3-chlorophenyl)propan-1-one