Order Entry
Ireland
ContactUsLinkComponent
 
Impurity and API standards, N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline
  PROOMM0378.06
 :  
Impurity and API standards, N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline
  PROOMM0378.06
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    51765-72-1
  • Description:
    N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline
  • Formula:
    C₁₄H₁₄N₂O₇S₂
  • Description:
    N,N-Bis(methylsulphonyl)-4-nitro-2-phenoxyaniline