Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, racecadotrils, Ph. Eur., RS
Organic reference standards, racecadotrils, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, racecadotrils, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    81110-69-2
  • Formula:
    C₁₉H₂₁NO₃S
  • Description:
    Racecadotril impurity G, benzyl [[(2RS)-2-benzyl-3-sulphanylpropanoyl]amino]acetate

Specifications

Frequently Bought Together