Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, promethazines, Ph. Eur., RS
Organic reference standards, promethazines, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, promethazines, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    7640-51-9
  • Formula:
    C₁₇H₂₀N₂OS
  • Description:
    Promethazine impurity D

Specifications

Frequently Bought Together