Impurity and API standard, phenylalanine
Catalog # PROOMM0314.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:150-30-1
- Description:Phenylalanine
- Formula:C₉H₁₁NO₂
- Description:Phenylalanine