0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Standards
Organic Standards
Impurity and API standard, prednienic acid
Impurity and API standard, prednienic acid
Impurity and API standard, prednienic acid
PROOMM0194.07
:
LGC Standards PROMOCHEM
:
Impurity and API standard, prednienic acid
PROOMM0194.07
:
LGC Standards PROMOCHEM
:
Pk:
100 mg
Standards:
Reference standards
CAS No.:
37927-29-0
Description:
Prednienic acid
Formula:
C₂₀H₂₆O₅
Description:
Prednienic Acid