Order Entry
Ireland
ContactUsLinkComponent
Impurity and API standard, losartan methyl ether
Impurity and API standard, losartan methyl ether
Catalog # PROOMM0168.12
CAS Number:  
Impurity and API standard, losartan methyl ether
Catalog # PROOMM0168.12
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    114798-94-6
  • Description:
    Losartan methyl ether
  • Formula:
    C₂₃H₂₅ClN₆O
  • Description:
    Losartan Methyl Ether

Specifications

About this item