Impurity and API standard, ketoprofen methyl ester
Catalog # PROOMM0001.32
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:47087-07-0
- Description:Methyl 2-(3-Benzoylphenyl)propionate
- Formula:C₁₇H₁₆O₃
- Description:Methyl 2-(3-Benzoylphenyl)propionate (Ketoprofen Methyl Ester)