Order Entry
Ireland
ContactUsLinkComponent
Organic reference standards, Moxifloxacin, Ph. Eur. RS
Organic reference standards, Moxifloxacin, Ph. Eur. RS
Catalog # EDQMY0000703
Supplier:  EDQM
CAS Number:  
Organic reference standards, Moxifloxacin, Ph. Eur. RS
Catalog # EDQMY0000703
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    120 mg
  • Standards:
    Reference standard
  • CAS No.:
    186826-86-8
  • Description:
    Moxifloxacin hydrochloride
  • Seq:
    01
  • Formula:
    C₂₁H₂₅ClFN₃O₄
  • Description:
    Moxifloxacin hydrochloride

Specifications

About this item