Connectors, tubing, Opta® SFT
Catalog # SART640FS014M-D
Supplier: Sartorius
Specifications
- Description:Connector, 1/4" hose barb, sterile, female small connector body
- Pk:10
Specifications
About this item
The Opta® SFT range of sterile connectors allows fast and reliable sterile connection and sterile fluid transfer between two separate, pre-sterilised process components in biopharmaceutical manufacturing operations.
- Easy, robust, repeatable operation
- Claimed size equivalent to tubing ID size with no flow restriction
- Safe and robust tubing connections
The OPTA® SFT-I connectors can be integrated into Sartorius Stedim Biotech Fluid Management Technologies assemblies and are also available as individual divise for end-user assembly with silicone tubing.