Organic reference standards, megestrol acetates, Ph. Eur.
: EDQM
- CAS No.:595-33-5
- Formula:C₂₄H₃₂O₄
- Description:Megestrol acetate
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection