Bottles, wide neck, clear, straight sided round
Supplier: BERLIN PACKAGING
Certificates
About this item
Made of glass, clear.
- Provide maximum content visibility
- Feature a wide opening
- Suggested as a general purpose bottle
Designed without shoulders for maximum storage capacity and content visibility.
Frequently Bought Together