Specifications
- Capacity:160 l
- Circulation pump (Qmax):Flow rate: 380 l/min
- Noise level:<63 dBa
- Temperature:95 °C
- Water consumption:13 l
- Description:Laboratory glassware washer/dryer, 910 LX, 380 V
- Seq:14
- Model:910 LX
- Chamber W×D×H:535×510×544 mm
- Cleaning mechanisms:Rotary dual spray arms located at the top and bottom of chamber
- Control:Microprocessor
- Controller/programs:Prolux controller/42
- Dosing of chemicals:Automatic
- Drainage:Drain pipe Ø 40 mm
- Drying power:Drying by direct injection of HEPA filtered air through injectors
- No. of washing levels:2
- Program operation/display:4 main wash programmes: chemistry glassware, bacteriology/virology (high temperature), stubborn stains (agar), volumetric glassware (low temperature) Visual and acoustic signal, indicator lamps for service and fault messages
- Water connection:Flexible coupling with female 3/4"
- Water heating power:9 kW
- Weight:143 kg
- W×D×H:604×736×1466 mm
- Pk:1
Specifications
About this item
The Lancer 910 LX offers 2 levels of cleaning with a choice of position for the upper level. This freestanding glassware washer while being contained in a small footprint, does not compromise on wash capacity. It includes a built-in drawer for the safe storage and input of chemicals. It also offers powerful forced air drying through the injectors and in the chamber to ensure complete and rapid drying of glassware both inside and out. Rack systems are common across the 810-1300 LX and 1800 LXA models enabling economical upgrade when expanding the capacity of your lab.
- Fast cycles and outstanding results from powerful wash action and design
- Easy to use microprocessor controls with self-diagnostic software and alarms
- Flexibility of multi-level injection washing and interchangeable racks
- All sizes and types of glassware cleaned with chamber configuration choices
- Ergonomic and safe loading of racks on load bearing telescopic runners
- Small footprint with surprisingly large capacity
- HEPA filtered chamber and direct injection drying reduces cycle time by drying glassware and labware inside and out
- Increased safety with on-board storage and input of chemicals
- Ergonomic door level height allows convenient loading and helps to avoid back strain
The 910 LX provides injection washing and drying on one or two levels simultaneously, delivering the wash/rinse solutions into narrow-necked glassware to provide for efficient and effective cleaning. Three washing positions are available and adjustable for the upper level and provide for greater wash load flexibility. Low profile, rear manifold connections provide wash/rinse solution to the racks, maximizing usable rack surface, which is often reduced by common centre manifold designs. 100% of the chamber volume is then available for an unbeaten washing capacity in this size/range of glassware washer dryer.
The 910 LX features microprocessor controls with 4 pre-set programs for chemistry glassware, bacteriology/virology (high temperature), stubborn stains (agar) and volumetric glassware (lower-temp). For other applications the 910 LX offers 38 additional programs that can be modified to fit any application. These custom programs offer the user the ability to easily adjust cycle parameters for program phases, temperatures, phase times and chemical dosing. The system control panel is user friendly with a 7 button keypad and 2 ½ inch LCD interface. Self-diagnostic software provides visual and audible alarms to quickly alert the user in the event of a malfunction, displaying the error and allowing the owner to avoid costly service calls.
With AISI 316 L stainless steel and electronically welded chamber (under Argon protection) and door, the unit is fully enclosed on all 6 sides with AISI 304 L stainless steel to protect components and wiring. Maximum durability of the washer components ensures a long life span. Additionally, the 910 LX is insulated with synthetic rubber based, closed cell foam to guard against heat lost and reduce noise levels creating lower utility costs and a better user experience in the lab.